Learn More
Abnova™ Human GNE Full-length ORF (NP_005467.1, 1 a.a. - 722 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_005467.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 10020 |
Molecular Weight (g/mol) | 105.7kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-946-123
|
Abnova™
H00010020P01S |
10 ug |
Each for $499.00
|
|
|||||
89-946-124
|
Abnova™
H00010020P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
The protein encoded by this gene is a bifunctional enzyme that initiates and regulates the biosynthesis of N-acetylneuraminic acid (NeuAc), a precursor of sialic acids. It is a rate-limiting enzyme in the sialic acid biosynthetic pathway. Sialic acid modification of cell surface molecules is crucial for their function in many biologic processes, including cell adhesion and signal transduction. Differential sialylation of cell surface molecules is also implicated in the tumorigenicity and metastatic behavior of malignant cells. Mutations in this gene are associated with sialuria, autosomal recessive inclusion body myopathy, and Nonaka myopathy. Alternative splicing of this gene results in transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MEKNGNNRKLRVCVATCNRADYSKLAPIMFGIKTEPEFFELDVVVLGSHLIDDYGNTYRMIEQDDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALALATSAALMNIRILHIEGGEVSGTIDDSIRHAITKLAHYHVCCTRSAEQHLISMCEDHDRILLAGCPSYDKLLSAKNKDYMSIIRMWLGDDVKSKDYIVALQHPVTTDIKHSIKMFELTLDALISFNKRTLVLFPNIDAGSKEMVRVMRKKGIEHHPNFRAVKHVPFDQFIQLVAHAGCMIGNSSCGVREVGAFGTPVINLGTRQIGRETGENVLHVRDADTQDKILQALHLQFGKQYPCSKIYGDGNAVPRILKFLKSIDLQEPLQKKFCFPPVKENISQDIDHILETLSALAVDLGGTNLRVAIVSMKGEIVKKYTQFNPKTYEERINLILQMCVEAAAEAVKLNCRILGVGISTGGRVNPREGIVLHSTKLIQEWNSVDLRTPLSDTLHLPVWVDNDGNCAALAERKFGQGKGLENFVTLITGTGIGGGIIHQHELIHGSSFCAAELGHLVVSLDGPDCSCGSHGCIEAYASGMALQREAKKLHDEDLLLVEGMSVPKDEAVGALHLIQAAKLGNAKAQSILRTAGTALGLGVVNILHTMNPSLVILSGVLASHYIHIVKDVIRQQALSSVQDVDVVVSDLVDPALLGAASMVLDYTTRRIYSpecifications
NP_005467.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
105.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEKNGNNRKLRVCVATCNRADYSKLAPIMFGIKTEPEFFELDVVVLGSHLIDDYGNTYRMIEQDDFDINTRLHTIVRGEDEAAMVESVGLALVKLPDVLNRLKPDIMIVHGDRFDALALATSAALMNIRILHIEGGEVSGTIDDSIRHAITKLAHYHVCCTRSAEQHLISMCEDHDRILLAGCPSYDKLLSAKNKDYMSIIRMWLGDDVKSKDYIVALQHPVTTDIKHSIKMFELTLDALISFNKRTLVLFPNIDAGSKEMVRVMRKKGIEHHPNFRAVKHVPFDQFIQLVAHAGCMIGNSSCGVREVGAFGTPVINLGTRQIGRETGENVLHVRDADTQDKILQALHLQFGKQYPCSKIYGDGNAVPRILKFLKSIDLQEPLQKKFCFPPVKENISQDIDHILETLSALAVDLGGTNLRVAIVSMKGEIVKKYTQFNPKTYEERINLILQMCVEAAAEAVKLNCRILGVGISTGGRVNPREGIVLHSTKLIQEWNSVDLRTPLSDTLHLPVWVDNDGNCAALAERKFGQGKGLENFVTLITGTGIGGGIIHQHELIHGSSFCAAELGHLVVSLDGPDCSCGSHGCIEAYASGMALQREAKKLHDEDLLLVEGMSVPKDEAVGALHLIQAAKLGNAKAQSILRTAGTALGLGVVNILHTMNPSLVILSGVLASHYIHIVKDVIRQQALSSVQDVDVVVSDLVDPALLGAASMVLDYTTRRIY | |
RUO | |
GNE | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
10020 | |
GNE (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DMRV/GLCNE/IBM2/NM/Uae1 | |
GNE | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.