Learn More
Abnova™ Human GOLPH3 Full-length ORF (NP_071413.1, 1 a.a. - 298 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_071413.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64083 |
Molecular Weight (g/mol) | 60.2kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-946-236
|
Abnova™
H00064083P01S |
10 ug |
Each for $499.00
|
|
|||||
89-946-237
|
Abnova™
H00064083P01L |
25 ug |
Each for $769.00
|
|
|||||
Description
The Golgi complex plays a key role in the sorting and modification of proteins exported from the endoplasmic reticulum. The protein encoded by this gene is a peripheral membrane protein of the Golgi stack and may have a regulatory role in Golgi trafficking. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of these variants has not been determined. [provided by RefSeq]
Sequence: MTSLTQRSSGLVQRRTEASRNAADKERAAGGGAGSSEDDAQSRRDEQDDDDKGDSKETRLTLMEEVLLLGLKDREGYTSFWNDCISSGLRGCMLIELALRGRLQLEACGMRRKSLLTRKVICKSDAPTGDVLLDEALKHVKETQPPETVQNWIELLSGETWNPLKLHYQLRNVRERLAKNLVEKGVLTTEKQNFLLFDMTTHPLTNNNIKQRLIKKVQEAVLDKWVNDPHRMDRRLLALIYLAHASDVLENAFAPLLDEQYDLATKRVRQLLDLDPEVECLKANTNEVLWAVVAAFTKSpecifications
NP_071413.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
60.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MTSLTQRSSGLVQRRTEASRNAADKERAAGGGAGSSEDDAQSRRDEQDDDDKGDSKETRLTLMEEVLLLGLKDREGYTSFWNDCISSGLRGCMLIELALRGRLQLEACGMRRKSLLTRKVICKSDAPTGDVLLDEALKHVKETQPPETVQNWIELLSGETWNPLKLHYQLRNVRERLAKNLVEKGVLTTEKQNFLLFDMTTHPLTNNNIKQRLIKKVQEAVLDKWVNDPHRMDRRLLALIYLAHASDVLENAFAPLLDEQYDLATKRVRQLLDLDPEVECLKANTNEVLWAVVAAFTK | |
RUO | |
GOLPH3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64083 | |
GOLPH3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ90675/GOPP1/GPP34/MIDAS | |
GOLPH3 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.