Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human Gpr65 Full-Length Orf (Np_003599.2) Recombinant Protein Without Tag.

Recombinant proteins expressed from in vitro wheat germ system. Such proteins mirror the conformation and folding of the native proteins in the eukaryotic biological system.

Manufacturer:  Abnova™ H00008477G01

Catalog No. 89-032-196


Add to cart

Description

Description

Good Folding

Immunogen Sequence: MNSTCIEEQHDLDHYLFPIVYIFVIIVSIPANIGSLCVSFLQAKKESELGIYLFSLSLSDLLYALTLPLWIDYTWNKDNWTFSPALCKGSAFLMYMNFYSSTAFLTCIAVDRYLAVVYPLKFFFLRTRRFALMVSLSIWILETIFNAVMLWEDETVVEYCDAEKSNFTLCYDKYPLEKWQINLNLFRTCTGYAIPLVTILICN

Specifications

Specifications

NP_003599.2
Liquid
8477
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
TDAG8|hTDAG8
GPR65
None
Antibody Production, Compound Screening, Functional Study
Liquid
GPR65 (Human) Recombinant Protein
MNSTCIEEQHDLDHYLFPIVYIFVIIVSIPANIGSLCVSFLQAKKESELGIYLFSLSLSDLLYALTLPLWIDYTWNKDNWTFSPALCKGSAFLMYMNFYSSTAFLTCIAVDRYLAVVYPLKFFFLRTRRFALMVSLSIWILETIFNAVMLWEDETVVEYCDAEKSNFTLCYDKYPLEKWQINLNLFRTCTGYAIPLVTILICNRKVYQAVRHNKATENKEKKRIIKLLVSITVTFVLCFTPFHVMLLIRCILEHAVNFEDHSNSGKRTYTMYRITVALTSLNCVADPILYCFVTETGRYDMWNILKFCTGRCNTSQRQRKRILSVSTKDTMELEVLE
RUO
GPR65
Wheat Germ (in vitro)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only