Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human GPSM1 Full-length ORF (AAH09979, 1 a.a. - 434 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | AAH09979 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 26086 |
Molecular Weight (g/mol) | 73.48kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-932-857
|
Abnova™
H00026086P01L |
25 ug |
Each for $769.00
|
|
|||||
89-932-856
|
Abnova™
H00026086P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
G proteins propagate intracellular signals initiated by G protein-coupled receptors. GPSM1, a receptor-independent activator of G protein signaling, is one of several factors that influence the basal activity of G protein signaling systems (Pizzinat et al., 2001 [PubMed 11278352]).[supplied by OMIM]
Sequence: MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQASpecifications
AAH09979 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
73.48kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEASCLELALEGERLCKAGDFKTGVAFFEAAVQVGTEDLKTLSAIYSQLGNAYFYLKEHGRALEYHKHDLLLARTIGDRMGEAKASGNLGNTLKVLGRFDEAAVCCQRHLSIAQEQGDKVGEARALYNIGNVYHAKGKQLSWNAANATQDPGHLPPDVRETLCKASEFYERNLSLVKELGDRAAQGRAYGNLGNTHYLLGNFTEATTFHKERLAIAKEFGDKAAERRAYSNLGNAHVFLGRFDVAAEYYKKTLQLSRQLRDQAVEAQACYSLGNTYTLLQDYERAAEYHLRHLLIAQELADRVGEGRACWSLGNAYVSMGRPAQALTFAKKHLQISQEIGDRHGELTARMNVAQLQLVLGRLTSPAASEKPDLAGYEAQGEFQGCGGVLLPTGTDRIRSCGGVGSRPGQHGGGGSRQKMAPTSQFFLASGTAQA | |
RUO | |
GPSM1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
26086 | |
GPSM1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AGS3/DKFZp727I051 | |
GPSM1 | |
Recombinant | |
wheat germ expression system |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title