Learn More
Raybiotech IncSupplier Diversity Partner Human GPV/CD42d ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHGPV1
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.31 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human GPV.
Catalog No. 50-194-9329