Learn More
Raybiotech Inc Human Granzyme B ELISA Kit
SDP
Supplier:  Raybiotech Inc ELHGZMB1
Encompass_Preferred
For serum, plasma, and cell culture supernatant samples. Sensitivity: 2.87 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Granzyme B. Other species not determined.
Catalog No. 50-194-9336