Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRHL2 Full-length ORF (NP_079191.1, 1 a.a. - 625 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00079977P01L

 View more versions of this product

Catalog No. 89-946-555


Add to cart

Description

Description

The protein encoded by this gene is a transcription factor that can act as a homodimer or as a heterodimer with either GRHL1 or GRHL3. Defects in this gene are a cause of non-syndromic sensorineural deafness autosomal dominant type 28 (DFNA28)

Sequence: MSQESDNNKRLVALVPMPSDPPFNTRRAYTSEDEAWKSYLENPLTAATKAMMSINGDEDSAAALGLLYDYYKVPRDKRLLSVSKASDSQEDQEKRNCLGTSEAQSNLSGGENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQKQGEGPMTYLNKGQFYAITLSETGDNKCFRHPISKVRSVVMVVFSEDKNRDEQLKYWKYWHSRQHTAKQRVLDIADYKESFNTIGNIEEIAYNAVSFTWDVNEEAKIFITVNCLSTDFSSQKGVKGLPLMIQIDTYSYNNRSNKPIHRAYCQIKVFCDKGAERKIRDEERKQNRKKGKGQASQTQCNSSSDGKLAAIPLQKKSDITYFKTMPDLHSQPVLFIPDVHFANLQRTGQVYYNTDDEREGGSVLVKRMFRPMEEEFGPVPSKQMKEEGTKRVLLYVRKETDDVFDALMLKSPTVKGLMEAISEKYGLPVEKIAKLYKKSKKGILVNMDDNIIEHYSNEDTFILNMESMVEGFKVTLMEI
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
97.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BOM|DFNA28|FLJ11172|FLJ13782|MGC149294|MGC149295|TFCP2L3
GRHL2
Yes
NP_079191.1
Liquid
79977
GRHL2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MSQESDNNKRLVALVPMPSDPPFNTRRAYTSEDEAWKSYLENPLTAATKAMMSINGDEDSAAALGLLYDYYKVPRDKRLLSVSKASDSQEDQEKRNCLGTSEAQSNLSGGENRVQVLKTVPVNLSLNQDHLENSKREQYSISFPESSAIIPVSGITVVKAEDFTPVFMAPPVHYPRGDGEEQRVVIFEQTQYDVPSLATHSAYLKDDQRSTPDSTYSESFKDAATEKFRSASVGAEEYMYDQTSSGTFQYTLEATKSLRQKQGEGPMTYLNKGQFYAITLSETGDNKCFRHPISKVRSVVMVVFSEDKNRDEQLKYWKYWHSRQHTAKQRVLDIADYKESFNTIGNIEEIAYNAVSFTWDVNEEAKIFITVNCLSTDFSSQKGVKGLPLMIQIDTYSYNNRSNKPIHRAYCQIKVFCDKGAERKIRDEERKQNRKKGKGQASQTQCNSSSDGKLAAIPLQKKSDITYFKTMPDLHSQPVLFIPDVHFANLQRTGQVYYNTDDEREGGSVLVKRMFRPMEEEFGPVPSKQMKEEGTKRVLLYVRKETDDVFDALMLKSPTVKGLMEAISEKYGLPVEKIAKLYKKSKKGILVNMDDNIIEHYSNEDTFILNMESMVEGFKVTLMEI
RUO
GRHL2
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit