Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRIK1 Partial ORF (NP_000821, 331 a.a. - 440 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002897Q01S

 View more versions of this product

Catalog No. 89-946-570


Add to cart

Description

Description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. The subunit encoded by this gene is subject to RNA editing (CAG->CGG; Q->R) within the second transmembrane domain, which is thought to alter the properties of ion flow. Alternative splicing, resulting in transcript variants encoding different isoforms, has been noted for this gene. [provided by RefSeq]

Sequence: VAIASHRASQLTVSSLQCHRHKPWRLGPRFMNLIKEARWDGLTGHITFNKTNGLRKDFDLDIISLKEEGTEKAAGEVSKHLYKVWKKIGIWNSNSGLNMTDSNKDKSSNI
Specifications

Specifications

NP_000821
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VAIASHRASQLTVSSLQCHRHKPWRLGPRFMNLIKEARWDGLTGHITFNKTNGLRKDFDLDIISLKEEGTEKAAGEVSKHLYKVWKKIGIWNSNSGLNMTDSNKDKSSNI
RUO
GRIK1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
2897
GRIK1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EAA3|EEA3|GLR5|GLUR5
GRIK1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit