Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRIK3 Full-length ORF (NP_000822.2, 1 a.a. - 919 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002899P01L

 View more versions of this product

Catalog No. 89-946-576


Add to cart

Description

Description

Glutamate receptors are the predominant excitatory neurotransmitter receptors in the mammalian brain and are activated in a variety of normal neurophysiologic processes. This gene product belongs to the kainate family of glutamate receptors, which are composed of four subunits and function as ligand-activated ion channels. It is not certain if the subunit encoded by this gene is subject to RNA editing as the other 2 family members (GRIK1 and GRIK2). A Ser310Ala polymorphism has been associated with schizophrenia, and there are conflicting reports of its association with the pathogenesis of delirium tremens in alcoholics. [provided by RefSeq]

Sequence: MTAPWRRLRSLVWEYWAGLLVCAFWIPDSRGMPHVIRIGGIFEYADGPNAQVMNAEEHAFRFSANIINRNRTLLPNTTLTYDIQRIHFHDSFEATKKACDQLALGVVAIFGPSQGSCTNAVQSICNALEVPHIQLRWKHHPLDNKDTFYVNLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGMMTEYYHFIFTTLDLYALDLEPYRYSGVNLTGFRILNVDNPHVSAIVEKWSMERLQAAPRSESGLLDGVMMTDAALLYDAVHIVSVCYQRAPQMTVNSLQCHRHKAWRFGGRFMNFIKEAQWEGLTGRIVFNKTSGLRTDFDLDIISLKEDGLEKVGVWSPADGLNITEVAKGRGPNVTDSLTNRSLIVTTVLEEPFVMFRKSDRTLYGNDRFEGYCIDLLKELAHILGFSYEIRLVEDGKYGAQDDKGQWNGMVKELIDHKADLAVAPLTITHVREKAIDFSKPFMTLGVSILYRKPNGTNPSVFSFLNPLSPDIWMYVLLAYLGVSCVLFVIARFSPYEWYDAHPCNPGSEVVENNFTLLNSFWFGMGSLMQQGSELMPKALSTRIIGGIWWFFTLIIISSYTANLAAFLTVERMESPIDSADDLAKQTKIEYGAVKDGATMTFFKKSKISTFEKMWAFMSSKPSALVKNNEEGIQRALTADYALLMESTTIEYVTQRNCNLTQIGGLIDSKGYGIGTPMGSPYRDKITIAILQLQEEDKLHIMKEKWWRGSGCPEEENKEASALGIQKIGGIFIVLAAGLVLSVLVAVGEFVYKLRKTAEREQRSFCSTVADEIRFSLTCQRRVKHKPQPPMMVKTDAVINMHTFNDRRLPGKDSMACSTSLAPVFP
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
130.4kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EAA5|GLR7|GLUR7|GluR7a
GRIK3
Yes
NP_000822.2
Liquid
2899
GRIK3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTAPWRRLRSLVWEYWAGLLVCAFWIPDSRGMPHVIRIGGIFEYADGPNAQVMNAEEHAFRFSANIINRNRTLLPNTTLTYDIQRIHFHDSFEATKKACDQLALGVVAIFGPSQGSCTNAVQSICNALEVPHIQLRWKHHPLDNKDTFYVNLYPDYASLSHAILDLVQYLKWRSATVVYDDSTGLIRLQELIMAPSRYNIRLKIRQLPIDSDDSRPLLKEMKRGREFRIIFDCSHTMAAQILKQAMAMGMMTEYYHFIFTTLDLYALDLEPYRYSGVNLTGFRILNVDNPHVSAIVEKWSMERLQAAPRSESGLLDGVMMTDAALLYDAVHIVSVCYQRAPQMTVNSLQCHRHKAWRFGGRFMNFIKEAQWEGLTGRIVFNKTSGLRTDFDLDIISLKEDGLEKVGVWSPADGLNITEVAKGRGPNVTDSLTNRSLIVTTVLEEPFVMFRKSDRTLYGNDRFEGYCIDLLKELAHILGFSYEIRLVEDGKYGAQDDKGQWNGMVKELIDHKADLAVAPLTITHVREKAIDFSKPFMTLGVSILYRKPNGTNPSVFSFLNPLSPDIWMYVLLAYLGVSCVLFVIARFSPYEWYDAHPCNPGSEVVENNFTLLNSFWFGMGSLMQQGSELMPKALSTRIIGGIWWFFTLIIISSYTANLAAFLTVERMESPIDSADDLAKQTKIEYGAVKDGATMTFFKKSKISTFEKMWAFMSSKPSALVKNNEEGIQRALTADYALLMESTTIEYVTQRNCNLTQIGGLIDSKGYGIGTPMGSPYRDKITIAILQLQEEDKLHIMKEKWWRGSGCPEEENKEASALGIQKIGGIFIVLAAGLVLSVLVAVGEFVYKLRKTAEREQRSFCSTVADEIRFSLTCQRRVKHKPQPPMMVKTDAVINMHTFNDRRLPGKDSMACSTSLAPVFP
RUO
GRIK3
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit