Learn More
Abnova™ Human GRIK4 Partial ORF (NP_055434, 21 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_055434 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2900 |
Molecular Weight (g/mol) | 37.84kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-946-578
|
Abnova™
H00002900Q01L |
25 ug |
Each for $769.00
|
|
|||||
89-946-577
|
Abnova™
H00002900Q01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene encodes a protein that belongs to the glutamate-gated ionic channel family. Glutamate functions as the major excitatory neurotransmitter in the central nervous system through activation of ligand-gated ion channels and G protein-coupled membrane receptors. The protein encoded by this gene forms functional heteromeric kainate-preferring ionic channels with the subunits encoded by related gene family members. [provided by RefSeq]
Sequence: SPHSLRIAAILDDPMECSRGERLSITLAKNRINRAPERLGKAKVEVDIFELLRDSEYETAETMCQILPKGVVAVLGPSSSPASSSIISNICGEKEVPHFKVAPEEFVKFQSpecifications
NP_055434 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
EAA1/GRIK/KA1 | |
GRIK4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2900 | |
GRIK4 (Human) Recombinant Protein (Q01) | |
SPHSLRIAAILDDPMECSRGERLSITLAKNRINRAPERLGKAKVEVDIFELLRDSEYETAETMCQILPKGVVAVLGPSSSPASSSIISNICGEKEVPHFKVAPEEFVKFQ | |
RUO | |
GRIK4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.