Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GRK6 Full-length ORF (AAH17272.1, 1 a.a. - 589 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002870P01

Catalog No. 89-946-610


Add to cart

Description

Description

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. Several transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MELENIVANTVLLKAREGGGGNRKGKSKKWRQMLQFPHISQCEELRLSLERDYHSLCERQPIGRLLFREFCATRPELSRCVAFLDGVAEYEVTPDDKRKACGRQLTQNFLSHTGPDLIPEVPRQLVTNCTQRLEQGPCKDLFQELTRLTHEYLSVAPFADYLDSIYFNRFLQWKWLERQPVTKNTFRQYRVLGKGGFGEVCACQVRATGKMYACKKLEKKRIKKRKGEAMALNEKQILEKVNSRFVVSLAYAYETKDALCLVLTLMNGGDLKFHIYHMGQAGFPEARAVFYAAEICCGLEDLHRERIVYRDLKPENILLDDHGHIRISDLGLAVHVPEGQTIKGRVGTVGYMAPEVVKNERYTFSPDWWALGCFLYEMIAGQSPFQQRKKKIKREEVERLVKEVPEEYSERFSPQARSLCSQLLCKDPAERLGCRGGSAREVKEHPLFKKLNFKRLGAGMLEPPFKPDPQAIYCKDVLDIEQFSTVKGVELEPTDQDFYQKFATGSVPIPWQNEMVETECFQELNVFGLDGSVPPDLDWKGQPPAPPKKGLLQRLFSRQRIAVETAATARKSSPPASSPQPEAPTSSWR
Specifications

Specifications

AAH17272.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
93.7kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32135|GPRK6
GRK6
Yes
wheat germ expression system
Liquid
2870
GRK6 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MELENIVANTVLLKAREGGGGNRKGKSKKWRQMLQFPHISQCEELRLSLERDYHSLCERQPIGRLLFREFCATRPELSRCVAFLDGVAEYEVTPDDKRKACGRQLTQNFLSHTGPDLIPEVPRQLVTNCTQRLEQGPCKDLFQELTRLTHEYLSVAPFADYLDSIYFNRFLQWKWLERQPVTKNTFRQYRVLGKGGFGEVCACQVRATGKMYACKKLEKKRIKKRKGEAMALNEKQILEKVNSRFVVSLAYAYETKDALCLVLTLMNGGDLKFHIYHMGQAGFPEARAVFYAAEICCGLEDLHRERIVYRDLKPENILLDDHGHIRISDLGLAVHVPEGQTIKGRVGTVGYMAPEVVKNERYTFSPDWWALGCFLYEMIAGQSPFQQRKKKIKREEVERLVKEVPEEYSERFSPQARSLCSQLLCKDPAERLGCRGGSAREVKEHPLFKKLNFKRLGAGMLEPPFKPDPQAIYCKDVLDIEQFSTVKGVELEPTDQDFYQKFATGSVPIPWQNEMVETECFQELNVFGLDGSVPPDLDWKGQPPAPPKKGLLQRLFSRQRIAVETAATARKSSPPASSPQPEAPTSSWR
RUO
GRK6
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit