Learn More
Abnova™ Human GSG2 (NP_114171.2, 1 a.a. - 798 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5558
Catalog No. 89-946-671
Add to cart
Specifications
Specifications
NP_114171.2
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
116kDa
Baculovirus infected insect cell (Sf21) expression system
73% by SDS-PAGE/CBB staining
5 ug
RUO
GSG2
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: Histone H3 peptide. ATP: 100 uM.
Yes
Baculovirus infected insect cell (Sf21) expression system
Liquid
83903
GSG2 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HASPIN
GSG2
Insect
GST