Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GSPT2 Full-length ORF (AAH36077.1, 1 a.a. - 628 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$460.50 - $709.50

Specifications

Name GSPT2 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 95.4kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-946-698
View Documents
Abnova™
H00023708P01S
10 ug
Each for $460.50
Add to cart
 
89-946-699
View Documents
Abnova™
H00023708P01L
25 ug
Each for $709.50
Add to cart
 
Description

Description

GSPT2 is closely related to GSPT1 (MIM 139259), a GTP-binding protein that plays an essential role at the G1- to S-phase transition of the cell cycle in yeast and human cells. GSPT1 is a positive regulator of translational accuracy and, in a binary complex with eRF1 (MIM 600285), functions as a polypeptide chain release factor.[supplied by OMIM]

Sequence: MDSGSSSSDSAPDCWDQVDMESTGSAPSGDGVSSAVAEAQREPLSSAFSRKLNVNAKPFVPNVHAAEFVPSFLRGPTQPPTLPAGSGSNDETCTGAGYPQGKRMGRGAPVEPSREEPLVSLEGSNSAVTMELSEPVVENGEVEMALEESWEHSKEVSEAEPGGGSSGDSGPPEESGQEMMEEKEEIRKSKSVIVPSGAPKKEHVNVVFIGHVDAGKSTIGGQIMFLTGMVDKRTLEKYEREAKEKNRETWYLSWALDTNQEERDKGKTVEVGRAYFETERKHFTILDAPGHKSFVPNMIGGASQADLAVLVISARKGEFETGFEKGGQTREHAMLAKTAGVKHLIVLINKMDDHTVNWSIERYEECKEKLVPFLKKVGFSPKKDIHFMPCSGLTGANIKEQSDFCPWYTGLPFIPYLDNLPNFNRSIDGPIRLPIVDKYKDMGTVVLGKLESGSIFKGQQLVMMPNKHNVEVLGILSDDTETDFVAPGENLKIRLKGIEEEEILPGFILCDPSNLCHSGRTFDVQIVIIEHKSIICPGYNAVLHIHTCIEEVEITALISLVDKKSGEKSKTRPRFVKQDQVCIARLRTAGTICLETFKDFPQMGRFTLRDEGKTIAIGKVLKLVQEKD
Specifications

Specifications

GSPT2 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
95.4kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit