Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GTF2I Full-length ORF (AAH04472.1, 36 a.a. - 274 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002969P01L

 View more versions of this product

Catalog No. 89-946-803


Add to cart

Description

Description

This gene encodes a multifunctional phosphoprotein with roles in transcription and signal transduction. It is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by the deletion of contiguous genes at chromosome 7q11.23. Alternative splicing results in multiple transcript variants. Related pseudogenes have been identified on chromosomes 7, 13 and 21. [provided by RefSeq]

Sequence: ELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGISFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGSHHSSEGNEGTEMEVPAEG
Specifications

Specifications

AAH04472.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.03kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
BAP-135|BAP135|BTKAP1|DIWS|FLJ38776|FLJ56355|IB291|SPIN|TFII-I|WBS|WBSCR6
GTF2I
Yes
wheat germ expression system
Liquid
2969
GTF2I (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
ELAKSKAEVACIAVYETDVFVVGTERGRAFVNTRKDFQKDFVKYCVEEEEKAAEMHKMKSTTQANRMSVDAVEIETLRKTVEDYFCFCYGKALGKSTVVPVPYEKMLRDQSAVVVQGLPEGVAFKHPENYDLATLKWILENKAGISFIIKRPFLEPKKHVGGRVMVTDADRSILSPGGSCGPIKVKTEPTEDSGISLEMAAVTVKEESEDPDYYQYNIQGSHHSSEGNEGTEMEVPAEG
RUO
GTF2I
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit