Learn More
Abnova™ Human GTF2IRD1 Partial ORF (NP_005676, 431 a.a. - 530 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009569Q01L
Description
The protein encoded by this gene contains five GTF2I-like repeats and each repeat possesses a potential helix-loop-helix (HLH) motif. It may have the ability to interact with other HLH-proteins and function as a transcription factor or as a positive transcriptional regulator under the control of Retinoblastoma protein. This gene is deleted in Williams-Beuren syndrome, a multisystem developmental disorder caused by deletion of multiple genes at 7q11.23. Alternative splicing of this gene generates at least 2 transcript variants. [provided by RefSeq]
Sequence: IFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSEDSpecifications
NP_005676 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
IFTGNKFTKDTTKLEPASPPEDTSAEVSRATVLDLAGNARSDKGSMSEDCGPGTSGELGGLRPIKIEPEDLDIIQVTVPDPSPTSEEMTDSMPGHLPSED | |
RUO | |
GTF2IRD1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9569 | |
GTF2IRD1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BEN/CREAM1/GTF3/MUSTRD1/RBAP2/WBS/WBSCR11/WBSCR12/hMusTRD1alpha1 | |
GTF2IRD1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.