Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human GUCY1A2 Partial ORF (NP_000846, 296 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00002977Q01S

 View more versions of this product

Catalog No. 89-946-866


Add to cart

Description

Description

Soluble guanylyl (or guanylate) cyclases are heterodimeric enzymes consisting of an alpha subunit, such as alpha-2 (GUCY1A2), and a beta subunit, typically beta-1 (GUCY1B3; MIM 139397), which are activated by nitric oxide (NO) and which catalyze conversion of GTP to 3-prime, 5-prime-cyclic GMP and pyrophosphate (Harteneck et al., 1991 [PubMed 1683630]).[supplied by OMIM]

Sequence: CENTNIMKNLPQGTSQVPADLRISINTFCRAFPFHLMFDPSMSVLQLGEGLRKQLRCDTHKVLKFEDCFEIVSPKVNATFERVLLRLSTPFVIRTKPEASGSENKD
Specifications

Specifications

NP_000846
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.4kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CENTNIMKNLPQGTSQVPADLRISINTFCRAFPFHLMFDPSMSVLQLGEGLRKQLRCDTHKVLKFEDCFEIVSPKVNATFERVLLRLSTPFVIRTKPEASGSENKD
RUO
GUCY1A2
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
2977
GUCY1A2 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GC-SA2|GUC1A2
GUCY1A2
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit