Learn More
Raybiotech IncSupplier Diversity Partner Human HAI-2 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELHHAI21
For cell culture supernatants, EDTA and/or Citrate treated plasma, and serum samples. Heparin is not recommended as an anticoagulant For use in this assay. Sensitivity: 3 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human HAI-2, less than 0.1% cross-reactivity with human HAI-1 and mouse HAI-2B is observed
Catalog No. 50-194-9339