Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HAO2 Full-length ORF (NP_001005783.1, 1 a.a. - 351 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$499.00 - $769.00

Specifications

Accession Number NP_001005783.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 51179
Molecular Weight (g/mol) 65.2kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-082
View Documents
Abnova™
H00051179P01L
25 ug
Each for $769.00
Add to Cart
 
89-947-081
View Documents
Abnova™
H00051179P01S
10 ug
Each for $499.00
Add to Cart
 
Description

Description

This gene is one of three related genes that have 2-hydroxyacid oxidase activity yet differ in encoded protein amino acid sequence, tissue expression and substrate preference. Subcellular location of the encoded protein is the peroxisome. Specifically, this gene is expressed predominantly in liver and kidney and has the highest activity toward the substrate 2-hydroxypalmitate. Two alternatively spliced variants encoding the same isoform have been described. [provided by RefSeq]

Sequence: MSLVCLTDFQAHAREQLSKSTRDFIEGGADDSITRDDNIAAFKRIRLRPRYLRDVSEVDTRTTIQGEEISAPICIAPTGFHCLVWPDGEMSTARAAQAAGICYITSTFASCSLEDIVIAAPEGLRWFQLYVHPDLQLNKQLIQRVESLGFKALVITLDTPVCGNRRHDIRNQLRRNLTLTDLQSPKKGNAIPYFQMTPISTSLCWNDLSWFQSITRLPIILKGILTKEDAELAVKHNVQGIIVSNHGGRQLDEVLASIDALTEVVAAVKGKIEVYLDGGVRTGNDVLKALALGAKCIFLGRPILWGLACKGEHGVKEVLNILTNEFHTSMALTGCRSVAEINRNLVQFSRL
Specifications

Specifications

NP_001005783.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.2kDa
Glutathione Sepharose 4 Fast Flow
MSLVCLTDFQAHAREQLSKSTRDFIEGGADDSITRDDNIAAFKRIRLRPRYLRDVSEVDTRTTIQGEEISAPICIAPTGFHCLVWPDGEMSTARAAQAAGICYITSTFASCSLEDIVIAAPEGLRWFQLYVHPDLQLNKQLIQRVESLGFKALVITLDTPVCGNRRHDIRNQLRRNLTLTDLQSPKKGNAIPYFQMTPISTSLCWNDLSWFQSITRLPIILKGILTKEDAELAVKHNVQGIIVSNHGGRQLDEVLASIDALTEVVAAVKGKIEVYLDGGVRTGNDVLKALALGAKCIFLGRPILWGLACKGEHGVKEVLNILTNEFHTSMALTGCRSVAEINRNLVQFSRL
RUO
HAO2
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
51179
HAO2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
GIG16/HAOX2
HAO2
Recombinant
wheat germ expression system
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit