Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HBS1L Full-length ORF (NP_006611.1, 1 a.a. - 684 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00010767P01S

 View more versions of this product

Catalog No. 89-947-126


Add to cart

Description

Description

This gene encodes a member of the GTP-binding elongation factor family. It is expressed in multiple tissues with the highest expression in heart and skeletal muscle. The intergenic region of this gene and the MYB gene has been identified to be a quantitative trait locus (QTL) controlling fetal hemoglobin level, and this region influnces erythrocyte, platelet, and monocyte counts as well as erythrocyte volume and hemoglobin content. DNA polymorphisms at this region associate with fetal hemoglobin levels and pain crises in sickle cell disease. A single nucleotide polymorphism in exon 1 of this gene is significantly associated with severity in beta-thalassemia/Hemoglobin E. Multiple alternatively spliced transcript variants encoding different protein isoforms have been found for this gene. [provided by RefSeq]

Sequence: MARHRNVRGYNYDEDFEDDDLYGQSVEDDYCISPSTAAQFIYSRRDKPSVEPVEEYDYEDLKESSNSVSNHQLSGFDQARLYSCLDHMREVLGDAVPDEILIEAVLKNKFDVQKALSGVLEQDRVQSLKDKNEATVSTGKIAKGKPVDSQTSRSESEIVPKVAKMTVSGKKQTMGFEVPGVSSEENGHSFHTPQKGPPIEDAIASSDVLETASKSANPPHTIQASEEQSSTPAPVKKSGKLRQQIDVKAELEKRQGGKQLLNLVVIGHVDAGKSTLMGHMLYLLGNINKRTMHKYEQESKKAGKASFAYAWVLDETGEERERGVTMDVGMTKFETTTKVITLMDAPGHKDFIPNMITGAAQADVAVLVVDASRGEFEAGFETGGQTREHGLLVRSLGVTQLAVAVNKMDQVNWQQERFQEITGKLGHFLKQAGFKESDVGFIPTSGLSGENLITRSQSSELTKWYKGLCLLEQIDSFKPPQRSIDKPFRLCVSDVFKDQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLHDEPVDWAAAGDHVSLTLVGMDIIKINVGCIFCGPKVPIKACTRFRARILIFNIEIPITKGFPVLLHYQTVSEPAVIKRLISVLNKSTGEVTKKKPKFLTKGQNALVELQTQRPIALELYKDFKELGRFMLRYGGSTIAAGVVTEIKE
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
101.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686L13262/EF-1a/ERFS/HBS1/HSPC276
HBS1L
Yes
NP_006611.1
Liquid
10767
HBS1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MARHRNVRGYNYDEDFEDDDLYGQSVEDDYCISPSTAAQFIYSRRDKPSVEPVEEYDYEDLKESSNSVSNHQLSGFDQARLYSCLDHMREVLGDAVPDEILIEAVLKNKFDVQKALSGVLEQDRVQSLKDKNEATVSTGKIAKGKPVDSQTSRSESEIVPKVAKMTVSGKKQTMGFEVPGVSSEENGHSFHTPQKGPPIEDAIASSDVLETASKSANPPHTIQASEEQSSTPAPVKKSGKLRQQIDVKAELEKRQGGKQLLNLVVIGHVDAGKSTLMGHMLYLLGNINKRTMHKYEQESKKAGKASFAYAWVLDETGEERERGVTMDVGMTKFETTTKVITLMDAPGHKDFIPNMITGAAQADVAVLVVDASRGEFEAGFETGGQTREHGLLVRSLGVTQLAVAVNKMDQVNWQQERFQEITGKLGHFLKQAGFKESDVGFIPTSGLSGENLITRSQSSELTKWYKGLCLLEQIDSFKPPQRSIDKPFRLCVSDVFKDQGSGFCITGKIEAGYIQTGDRLLAMPPNETCTVKGITLHDEPVDWAAAGDHVSLTLVGMDIIKINVGCIFCGPKVPIKACTRFRARILIFNIEIPITKGFPVLLHYQTVSEPAVIKRLISVLNKSTGEVTKKKPKFLTKGQNALVELQTQRPIALELYKDFKELGRFMLRYGGSTIAAGVVTEIKE
RUO
HBS1L
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit