Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HCFC1 Full-length ORF (AAH63435.1, 1 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003054P01S

 View more versions of this product

Catalog No. 89-947-150

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene is a member of the host cell factor family and encodes a protein with five Kelch repeats, a fibronectin-like motif, and six HCF repeats, each of which contains a highly specific cleavage signal. This nuclear coactivator is proteolytically cleaved at one of the six possible sites, resulting in the creation of an N-terminal chain and the corresponding C-terminal chain. The final form of this protein consists of noncovalently bound N- and C-terminal chains. The protein is involved in control of the cell cycle and transcriptional regulation during herpes simplex virus infection. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Sequence: MASAVSPANLPAVLLQPRWKRVVGWSGPVPRPRHGHRAVAIKELIVVFGGGNEGIVDELHVYNTATNQWFIPAVRGDIPPGCAAYGFVCDGTRLLVFGGMVEYGKYSNDLYELQASRWEWKRLKAKTPKNGPPPCPRLGHSFSLVGNKCYLFGGLANDSEDPKNNIPRYLNDLYILELRPGSGVVAWDIPITYGVLPPPRESHTAVVYTEKDNKKSKLVIYGGMSGCRLGDLWTLDIDTLTWNKPSLSGVAPLPRSLHSATTIGNKMYVFGGWVPLVMDDVKVATHEKEWKCTNTLACLNLDTMAWETILMDTLEDNIPRARAGHCAVAINTRLYIWSGRDGYRKAWNNQVCCKDLWYLETEKPPPPARVQLVRANTNSLEVSWGAVATADSYLLQLQKYDIPATAATATSPTPNPVPSVPANPPKSL
Specifications

Specifications

AAH63435.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
73.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CFF|HCF-1|HCF1|HFC1|MGC70925|VCAF
HCFC1
Yes
wheat germ expression system
Liquid
3054
HCFC1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MASAVSPANLPAVLLQPRWKRVVGWSGPVPRPRHGHRAVAIKELIVVFGGGNEGIVDELHVYNTATNQWFIPAVRGDIPPGCAAYGFVCDGTRLLVFGGMVEYGKYSNDLYELQASRWEWKRLKAKTPKNGPPPCPRLGHSFSLVGNKCYLFGGLANDSEDPKNNIPRYLNDLYILELRPGSGVVAWDIPITYGVLPPPRESHTAVVYTEKDNKKSKLVIYGGMSGCRLGDLWTLDIDTLTWNKPSLSGVAPLPRSLHSATTIGNKMYVFGGWVPLVMDDVKVATHEKEWKCTNTLACLNLDTMAWETILMDTLEDNIPRARAGHCAVAINTRLYIWSGRDGYRKAWNNQVCCKDLWYLETEKPPPPARVQLVRANTNSLEVSWGAVATADSYLLQLQKYDIPATAATATSPTPNPVPSVPANPPKSL
RUO
HCFC1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit