Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HCFC1 Full-length ORF (AAH63435.1, 1 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name HCFC1 (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 73.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-151 View Documents Abnova™
H00003054P01L
25 ug Each for $599.28
Add to cart
 
89-947-150 View Documents Abnova™
H00003054P01S
10 ug Each for $384.10
Add to cart
 
Description

Description

This gene is a member of the host cell factor family and encodes a protein with five Kelch repeats, a fibronectin-like motif, and six HCF repeats, each of which contains a highly specific cleavage signal. This nuclear coactivator is proteolytically cleaved at one of the six possible sites, resulting in the creation of an N-terminal chain and the corresponding C-terminal chain. The final form of this protein consists of noncovalently bound N- and C-terminal chains. The protein is involved in control of the cell cycle and transcriptional regulation during herpes simplex virus infection. Alternatively spliced variants which encode different protein isoforms have been described; however, not all variants have been fully characterized. [provided by RefSeq]

Sequence: MASAVSPANLPAVLLQPRWKRVVGWSGPVPRPRHGHRAVAIKELIVVFGGGNEGIVDELHVYNTATNQWFIPAVRGDIPPGCAAYGFVCDGTRLLVFGGMVEYGKYSNDLYELQASRWEWKRLKAKTPKNGPPPCPRLGHSFSLVGNKCYLFGGLANDSEDPKNNIPRYLNDLYILELRPGSGVVAWDIPITYGVLPPPRESHTAVVYTEKDNKKSKLVIYGGMSGCRLGDLWTLDIDTLTWNKPSLSGVAPLPRSLHSATTIGNKMYVFGGWVPLVMDDVKVATHEKEWKCTNTLACLNLDTMAWETILMDTLEDNIPRARAGHCAVAINTRLYIWSGRDGYRKAWNNQVCCKDLWYLETEKPPPPARVQLVRANTNSLEVSWGAVATADSYLLQLQKYDIPATAATATSPTPNPVPSVPANPPKSL
Specifications

Specifications

HCFC1 (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
73.6kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit