Learn More
Abnova™ Human HCK (PO8631, 230 a.a. - 947 a.a.) Partial Recombinant Protein expressed in Sf9 cells
Used for Func, SDS-PAGE
Supplier: Abnova™ P4204
Description
The protein encoded by this gene is a protein-tyrosine kinase that is predominantly expressed in hemopoietic cell types. The encoded protein may help couple the Fc receptor to the activation of the respiratory burst. In addition, it may play a role in neutrophil migration and in the degranulation of neutrophils. Alternate translation initiation site usage, including a non-AUG (CUG) codon, results in the production of two different isoforms, that have different subcellular localization. [provided by RefSeq]
Specifications
PO8631 | |
Liquid | |
57kDa | |
Insect cell (Sf9) expression system | |
5 ug | |
RUO | |
HCK | |
171nmol phosphate incorporated into poly(Glu-Tyr) per minute per mg protein at 30°C for 15 minutes using a final concentration of 50μM ATP (0.83 uCi/assay). | |
Recombinant | |
Insect cell (Sf9) expression system | |
≥90% by SDS-PAGE |
Functional Study, SDS-PAGE | |
3055 | |
HCK (Human) Recombinant Protein | |
1 ug/lane SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
JTK9 | |
HCK | |
Baculovirus | |
None | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.