Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HEBP1 Partial ORF (NP_057071, 80 a.a. - 189 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00050865Q01S

 View more versions of this product

Catalog No. 89-947-245


Add to cart

Description

Description

The full-length protein encoded by this gene is an intracellular tetrapyrrole-binding protein. This protein includes a natural chemoattractant peptide of 21 amino acids at the N-terminus, which is a natural ligand for formyl peptide receptor-like receptor 2 (FPRL2) and promotes calcium mobilization and chemotaxis in monocytes and dendritic cells. [provided by RefSeq]

Sequence: VPISFAVFPNEDGSLQKKLKVWFRIPNQFQSDPPAPSDKSVKIEEREGITVYSMQFGGYAKEADYVAQATRLRAALEGTATYRGDIYFCTGYDPPMKPYGRRNEIWLLKT
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VPISFAVFPNEDGSLQKKLKVWFRIPNQFQSDPPAPSDKSVKIEEREGITVYSMQFGGYAKEADYVAQATRLRAALEGTATYRGDIYFCTGYDPPMKPYGRRNEIWLLKT
RUO
HEBP1
Wheat Germ (in vitro)
GST
NP_057071
Liquid
50865
HEBP1 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HBP/HEBP
HEBP1
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit