Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HECA Partial ORF (NP_057301, 434 a.a. - 543 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051696Q01S

 View more versions of this product

Catalog No. 89-947-255


Add to cart

Description

Description

This gene encodes the homolog of the Drosophila headcase protein, a highly basic, cytoplasmic protein that regulates the re-entry of imaginal cells into the mitotic cycle during adult morphogenesis. In Drosophila, the encoded protein also inhibits terminal branching of neighboring cells during tracheal development. [provided by RefSeq]

Sequence: CHLQGRLMHLYAVCVDCLEGVHKIICIKCKSRWDGSWHQLGTMYTYDILAASPCCQARLNCKHCGKPVIDVRIGMQYFSEYSNVQQCPHCGNLDYHFVKPFSSFKVLEAY
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
CHLQGRLMHLYAVCVDCLEGVHKIICIKCKSRWDGSWHQLGTMYTYDILAASPCCQARLNCKHCGKPVIDVRIGMQYFSEYSNVQQCPHCGNLDYHFVKPFSSFKVLEAY
RUO
HECA
Wheat Germ (in vitro)
GST
NP_057301
Liquid
51696
HECA (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HDC/HDCL/HHDC/dJ225E12.1
HECA
Yes
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit