Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HEY2 Full-length ORF (NP_036391.1, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00023493P01S

 View more versions of this product

Catalog No. 89-947-336


Add to cart

Description

Description

This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The encoded protein forms homo- or hetero-dimers that localize to the nucleus and interact with a histone deacetylase complex to repress transcription. Expression of this gene is induced by the Notch signal transduction pathway. Two similar and redundant genes in mouse are required for embryonic cardiovascular development, and are also implicated in neurogenesis and somitogenesis. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. [provided by RefSeq]

Sequence: MKRPCEETTSESDMDETIDVGSENNYSGQSTSSVIRLNSPTTTSQIMARKKRRGIIEKRRRDRINNSLSELRRLVPTAFEKQGSAKLEKAEILQMTVDHLKMLQATGGKGYFDAHALAMDFMSIGFRECLTEVARYLSSVEGLDSSDPLRVRLVSHLSTCATQREAAAMTSSMAHHHHPLHPHHWAAAFHHLPAALLQPNGLHASESTPCRLSTTSEVPPAHGSALLTATFAHADSALRMPSTGSVAPCVPPLSTSLLSLSATVHAAAAAATAAAHSFPLSFAGAFPMLPPNAAAAVAAATAISPPLSVSATSSPQQTSSGTNNKPYRPWGTEVGAF
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.2kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CHF1/GRIDLOCK/GRL/HERP1/HESR2/HRT2/MGC10720/bHLHb32
HEY2
Yes
NP_036391.1
Liquid
23493
HEY2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MKRPCEETTSESDMDETIDVGSENNYSGQSTSSVIRLNSPTTTSQIMARKKRRGIIEKRRRDRINNSLSELRRLVPTAFEKQGSAKLEKAEILQMTVDHLKMLQATGGKGYFDAHALAMDFMSIGFRECLTEVARYLSSVEGLDSSDPLRVRLVSHLSTCATQREAAAMTSSMAHHHHPLHPHHWAAAFHHLPAALLQPNGLHASESTPCRLSTTSEVPPAHGSALLTATFAHADSALRMPSTGSVAPCVPPLSTSLLSLSATVHAAAAAATAAAHSFPLSFAGAFPMLPPNAAAAVAAATAISPPLSVSATSSPQQTSSGTNNKPYRPWGTEVGAF
RUO
HEY2
Wheat Germ (in vitro)
GST
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit