Learn More
Abnova™ Human HEY2 Full-length ORF (NP_036391.1, 1 a.a. - 337 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00023493P01L
Description
This gene encodes a member of the hairy and enhancer of split-related (HESR) family of basic helix-loop-helix (bHLH)-type transcription factors. The encoded protein forms homo- or hetero-dimers that localize to the nucleus and interact with a histone deacetylase complex to repress transcription. Expression of this gene is induced by the Notch signal transduction pathway. Two similar and redundant genes in mouse are required for embryonic cardiovascular development, and are also implicated in neurogenesis and somitogenesis. Alternatively spliced transcript variants have been found, but their biological validity has not been determined. [provided by RefSeq]
Sequence: MKRPCEETTSESDMDETIDVGSENNYSGQSTSSVIRLNSPTTTSQIMARKKRRGIIEKRRRDRINNSLSELRRLVPTAFEKQGSAKLEKAEILQMTVDHLKMLQATGGKGYFDAHALAMDFMSIGFRECLTEVARYLSSVEGLDSSDPLRVRLVSHLSTCATQREAAAMTSSMAHHHHPLHPHHWAAAFHHLPAALLQPNGLHASESTPCRLSTTSEVPPAHGSALLTATFAHADSALRMPSTGSVAPCVPPLSTSLLSLSATVHAAAAAATAAAHSFPLSFAGAFPMLPPNAAAAVAAATAISPPLSVSATSSPQQTSSGTNNKPYRPWGTEVGAFSpecifications
NP_036391.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
62.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CHF1/GRIDLOCK/GRL/HERP1/HESR2/HRT2/MGC10720/bHLHb32 | |
HEY2 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
23493 | |
HEY2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MKRPCEETTSESDMDETIDVGSENNYSGQSTSSVIRLNSPTTTSQIMARKKRRGIIEKRRRDRINNSLSELRRLVPTAFEKQGSAKLEKAEILQMTVDHLKMLQATGGKGYFDAHALAMDFMSIGFRECLTEVARYLSSVEGLDSSDPLRVRLVSHLSTCATQREAAAMTSSMAHHHHPLHPHHWAAAFHHLPAALLQPNGLHASESTPCRLSTTSEVPPAHGSALLTATFAHADSALRMPSTGSVAPCVPPLSTSLLSLSATVHAAAAAATAAAHSFPLSFAGAFPMLPPNAAAAVAAATAISPPLSVSATSSPQQTSSGTNNKPYRPWGTEVGAF | |
RUO | |
HEY2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.