Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HGF Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems AB294NA
This item is not returnable.
View return policy
Description
HGF Polyclonal specifically detects HGF in Human samples. It is validated for Western Blot, Neutralization.Specifications
HGF | |
Polyclonal | |
Unconjugated | |
P14210 | |
HGF | |
S. frugiperda insect ovarian cell line Sf 21-derived recombinant human HGF (R&D Systems, Catalog # 294-HG) Gln32-Ser728 Accession # P14210 | |
1 mg | |
Primary | |
Detects human HGF in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse HGF is observed. | |
Human | |
Purified |
Western Blot, Neutralization | |
LYOPH | |
Western Blot 1 ug/mL, Neutralization <2 ug/mL | |
deafness, autosomal recessive 39, DFNB39, EC 3.4.21, EC 3.4.21.7, fibroblast-derived tumor cytotoxic factor, F-TCF, hepatocyte growth factor (hepapoietin A; scatter factor), Hepatopoeitin-A, Hepatopoietin A, HGFB, HPTA, HPTAhepatocyte growth factor, lung fibroblast-derived mitogen, Scatter factor, SF, SFhepatopoeitin-A | |
Goat | |
Protein A or G purified | |
RUO | |
3082 | |
Reconstitute at 1 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction