Learn More
Abnova™ Human HGF (heterodimer) (P14210, alpha chain 463 amino acids, beta chain 234 amino acids) Recombinant Protein expressed in Hi-5 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P4085
Catalog No. 89-947-359
Add to cart
Description
Description
Hepatocyte growth factor regulates cell growth, cell motility, and morphogenesis by activating a tyrosine kinase signaling cascade after binding to the proto-oncogenic c-Met receptor. Hepatocyte growth factor is secreted by mesenchymal cells and acts as a multi-functional cytokine on cells of mainly epithelial origin. Its ability to stimulate mitogenesis, cell motility, and matrix invasion gives it a central role in angiogenesis, tumorogenesis, and tissue regeneration. It is secreted as a single inactive polypeptide and is cleaved by serine proteases into a 69-kDa alpha-chain and 34-kDa beta-chain. A disulfide bond between the alpha and beta chains produces the active, heterodimeric molecule. The protein belongs to the plasminogen subfamily of S1 peptidases but has no detectable protease activity. Alternative splicing of this gene produces multiple transcript variants encoding different isoforms. [provided by RefSeq]
Specifications
Specifications
P14210
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Lyophilized
78kDa
Insect cell (Hi-5) expression system
10 ug
RUO
F-TCF|HGFB|HPTA|SF
HGF
Insect
None
Insect cell (Hi-5) expression system
Lyophilized
3082
HGF (Human) Recombinant Protein
≥98% by SDS-PAGE and HPLC
Store at -20°C on dry atmosphere. After reconstitution with sterilized water, store at -20°C. Aliquot to avoid repeated freezing and thawing.
<0.1ng/μg
HGF
The ED50 determined by the scattering activity in the MDCK cell assay is typically at 0.5-1.0ng/mL.
Yes