Learn More
R&D Systems™ Human HGF Quantikine ELISA Kit, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.
a 4.25 or 4.5 hour solid phase ELISA that is designed to measure HGF levels in cell culture supernates, serum, and plasma.
Manufacturer:  R&D Systems™ SHG00
 View more versions of this product
Catalog No. SHG00
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
It contains Sf 21-expressed recombinant human pro-HGF and antibodies raised against the recombinant factor and has been shown to quantitate recombinant human HGF accurately. Results obtained using natural HGF showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural HGF
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates, Serum, EDTA Plasma, Citrate Plasma
Human
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules
3082
6 Plates
40pg/mL
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Citrate Plasma (50μL)
Sufficient materials to run ELISAs on at least six 96-well plates