Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HGSNAT Full-length ORF (AAH12452.1, 1 a.a. - 206 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00138050P01

Catalog No. 89-947-368


Add to cart

Description

Description

This gene encodes a lysosomal acetyltransferase, which is one of several enzymes involved in the lysosomal degradation of heparin sulfate. Mutations in this gene are associated with Sanfilippo syndrome C, one type of the lysosomal storage disease mucopolysaccaridosis III, which results from impaired degradation of heparan sulfate. [provided by RefSeq]

Sequence: MALGLCRCFHPRHSMAAFGLFPALPSALNSHPACTCLLDPSTWRPAHVSGPALASSPQILSVFSLGFPGFVNGSCVSRYKPDIIFPPGLPPPDLPSSVSIFYLQLLCSHGHCCITESGPLLSFSNWPPSLVPHFLKSPVHCHQIKLSPARSPLSEKPPLTWKHHCLAHILTYSPSRLDPHTSFQPPLPLHSLLPPPPPHPLVSPPL
Specifications

Specifications

AAH12452.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
48.7kDa
Glutathione Sepharose 4 Fast Flow
2 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686G24175|FLJ22242|FLJ32731|HGNAT|MPS3C|TMEM76
HGSNAT
Yes
wheat germ expression system
Liquid
138050
HGSNAT (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MALGLCRCFHPRHSMAAFGLFPALPSALNSHPACTCLLDPSTWRPAHVSGPALASSPQILSVFSLGFPGFVNGSCVSRYKPDIIFPPGLPPPDLPSSVSIFYLQLLCSHGHCCITESGPLLSFSNWPPSLVPHFLKSPVHCHQIKLSPARSPLSEKPPLTWKHHCLAHILTYSPSRLDPHTSFQPPLPLHSLLPPPPPHPLVSPPL
RUO
HGSNAT
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit