Learn More
Abnova™ Human HIPK4 (NP_653286.2, 1 a.a. - 616 a.a.) Full-length Recombinant Protein with GST-tag expressed in baculovirus infected Sf21 cells
Used for Func, SDS-PAGE
Manufacturer:  Abnova™ P5562
Catalog No. 89-947-429
Add to cart
Specifications
Specifications
Baculovirus infected insect cell (Sf21) expression system
Functional Study, Sodium Dodecyl Sulfate–Polyacrylamide Gel Electrophoresis
Liquid
96kDa
Baculovirus infected insect cell (Sf21) expression system
70% by SDS-PAGE/CBB staining
5 ug
RUO
HIPK4
The activity was measured by off-chip mobility shift assay. The enzyme was incubated with fluorescence-labeled substrate and Mg(or Mn)/ATP. The phosphorylated and unphosphorylated substrates were separated and detected by LabChip 3000. Substrate: DYRKtide-F. ATP: 100 uM.
Yes
NP_653286.2
Liquid
147746
HIPK4 (Human) Recombinant Protein
Glutathione sepharose chromatography
Loading 1 ug protein in SDS-PAGE
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ32818
HIPK4
Insect
GST