Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ Human HLCS Full-length ORF (NP_000402.3, 1 a.a. - 726 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00003141P01S
Description
Holocarboxylase synthetase (EC 6.3.4.10) covalently links biotin to propionyl-CoA carboxylase (PCCA; MIM 232000), pyruvate carboxylase (PC; MIM 608786), alpha-methylcrotonyl-CoA carboxylase (MCCC1; MIM 609010), and acetyl-CoA carboxylase (ACACA; MIM 200350).[supplied by OMIM]
Sequence: MEDRLHMDNGLVPQKIVSVHLQDSTLKEVKDQVSNKQAQILEPKPEPSLEIKPEQDGMEHVGRDDPKALGEEPKQRRGSASGSEPAGDSDRGGGPVEHYHLHLSSCHECLELENSTIESVKFASAENIPDLPYDYSSSLESVADETSPEREGRRVNLTGKAPNILLYVGSDSQEALGRFHEVRSVLADCVDIDSYILYHLLEDSALRDPWTDNCLLLVIATRESIPEDLYQKFMAYLSQGGKVLGLSSSFTFGGFQVTSKGALHKTVQNLVFSKADQSEVKLSVLSSGCRYQEGPVRLSPGRLQGHLENEDKDRMIVHVPFGTRGGEAVLCQVHLELPPSSNIVQTPEDFNLLKSSNFRRYEVLREILTTLGLSCDMKQVPALTPLYLLSAAEEIRDPLMQWLGKHVDSEGEIKSGQLSLRFVSSYVSEVEITPSCIPVVTNMEAFSSEHFNLEIYRQNLQTKQLGKVILFAEVTPTTMRLLDGLMFQTPQEMGLIVIAARQTEGKGRGGNVWLSPVGCALSTLLISIPLRSQLGQRIPFVQHLMSVAVVEAVRSIPEYQDINLRVKWPNDIYYSDLMKIGGVLVNSTLMGETFYILIGCGFNVTNSNPTICINDLITEYNKQHKAELKPLRADYLIARVVTVLEKLIKEFQDKGPNSVLPLYYRYWVHSGQQVHLGSAEGPKVSIVGLDDSGFLQVHQEGGEVVTVHPDGNSFDMLRNLILPKRRSpecifications
NP_000402.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HCS | |
HLCS | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
3141 | |
HLCS (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEDRLHMDNGLVPQKIVSVHLQDSTLKEVKDQVSNKQAQILEPKPEPSLEIKPEQDGMEHVGRDDPKALGEEPKQRRGSASGSEPAGDSDRGGGPVEHYHLHLSSCHECLELENSTIESVKFASAENIPDLPYDYSSSLESVADETSPEREGRRVNLTGKAPNILLYVGSDSQEALGRFHEVRSVLADCVDIDSYILYHLLEDSALRDPWTDNCLLLVIATRESIPEDLYQKFMAYLSQGGKVLGLSSSFTFGGFQVTSKGALHKTVQNLVFSKADQSEVKLSVLSSGCRYQEGPVRLSPGRLQGHLENEDKDRMIVHVPFGTRGGEAVLCQVHLELPPSSNIVQTPEDFNLLKSSNFRRYEVLREILTTLGLSCDMKQVPALTPLYLLSAAEEIRDPLMQWLGKHVDSEGEIKSGQLSLRFVSSYVSEVEITPSCIPVVTNMEAFSSEHFNLEIYRQNLQTKQLGKVILFAEVTPTTMRLLDGLMFQTPQEMGLIVIAARQTEGKGRGGNVWLSPVGCALSTLLISIPLRSQLGQRIPFVQHLMSVAVVEAVRSIPEYQDINLRVKWPNDIYYSDLMKIGGVLVNSTLMGETFYILIGCGFNVTNSNPTICINDLITEYNKQHKAELKPLRADYLIARVVTVLEKLIKEFQDKGPNSVLPLYYRYWVHSGQQVHLGSAEGPKVSIVGLDDSGFLQVHQEGGEVVTVHPDGNSFDMLRNLILPKRR | |
RUO | |
HLCS | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction