Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HMGCS2 Partial ORF (NP_005509.1, 424 a.a. - 508 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name HMGCS2 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 35.09kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-638 View Documents Abnova™
H00003158Q01L
25 ug Each for $599.28
Add to cart
 
89-947-637 View Documents Abnova™
H00003158Q01S
10 ug Each for $384.10
Add to cart
 
Description

Description

Mitochondrial 3-hydroxy-3-methylglutaryl CoA synthase (HMGCS2; EC 2.3.3.10) mediates the first reaction of ketogenesis, a metabolic pathway that provides lipid-derived energy for brain, heart, kidney, and other organs during times of carbohydrate deprivation such as fasting (Robinson and Williamson, 1980 [PubMed 6986618]). Also see cytoplasmic HMG-CoA synthase (HMGCS1; MIM 142940), which mediates an early step in cholesterol synthesis.[supplied by OMIM]

Sequence: RVSQDAAPGSPLDKLVSSTSDLPKRLASRKCVSPEEFTEIMNQREQFYHKVNFSPPGDTNSLFPGTWYLERVDEQHRRKYARRPV
Specifications

Specifications

HMGCS2 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
35.09kDa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit