Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HNRPF Full-length ORF (NP_004957.1, 1 a.a. - 415 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003185P01L

 View more versions of this product

Catalog No. 89-947-682


Add to cart

Description

Description

This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA binding proteins that complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and regulate alternative splicing, polyadenylation, and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has three repeats of quasi-RRM domains that bind to RNAs which have guanosine-rich sequences. This protein is very similar to the family member hnRPH. Multiple alternatively spliced variants, encoding the same protein, have been identified. [provided by RefSeq]

Sequence: MMLGPEGGEGFVVKLRGLPWSCSVEDVQNFLSDCTIHDGAAGVHFIYTREGRQSGEAFVELGSEDDVKMALKKDRESMGHRYIEVFKSHRTEMDWVLKHSGPNSADSANDGFVRLRGLPFGCTKEEIVQFFSGLEIVPNGITLPVDPEGKITGEAFVQFASQELAEKALGKHKERIGHRYIEVFKSSQEEVRSYSDPPLKFMSVQRPGPYDRPGTARRYIGIVKQAGLERMRPGAYSTGYGGYEEYSGLSDGYGFTTDLFGRDLSYCLSGMYDHRYGDSEFTVQSTTGHCVHMRGLPYKATENDIYNFFSPLNPVRVHIEIGPDGRVTGEADVEFATHEEAVAAMSKDRANMQHRYIELFLNSTTGASNGAYSSQVMQGMGVSAAQATYSGLESQSVSGCYGAGYSGQNSMGGYD
Specifications

Specifications

wheat germ expression system
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
72.1kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HNRPF|MGC110997|OK/SW-cl.23|mcs94-1
HNRNPF
Yes
NP_004957.1
Liquid
3185
HNRPF (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MMLGPEGGEGFVVKLRGLPWSCSVEDVQNFLSDCTIHDGAAGVHFIYTREGRQSGEAFVELGSEDDVKMALKKDRESMGHRYIEVFKSHRTEMDWVLKHSGPNSADSANDGFVRLRGLPFGCTKEEIVQFFSGLEIVPNGITLPVDPEGKITGEAFVQFASQELAEKALGKHKERIGHRYIEVFKSSQEEVRSYSDPPLKFMSVQRPGPYDRPGTARRYIGIVKQAGLERMRPGAYSTGYGGYEEYSGLSDGYGFTTDLFGRDLSYCLSGMYDHRYGDSEFTVQSTTGHCVHMRGLPYKATENDIYNFFSPLNPVRVHIEIGPDGRVTGEADVEFATHEEAVAAMSKDRANMQHRYIELFLNSTTGASNGAYSSQVMQGMGVSAAQATYSGLESQSVSGCYGAGYSGQNSMGGYD
RUO
HNRNPF
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit