Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOM-TES-103 Full-length ORF (NP_542769.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00025900P01S

 View more versions of this product

Catalog No. 89-947-708


Add to cart

Description

Description

This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Alternative splicing has been observed for this gene and three transcript variants encoding different isoforms have been identified. Other alternatively spliced transcripts may exist, but their biological validity has not been confirmed. [provided by RefSeq]

Sequence: MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMR
Specifications

Specifications

NP_542769.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp586I2223|FLJ20703|HOM-TES-103|IFFO|MGC117359
IFFO1
Yes
wheat germ expression system
Liquid
25900
HOM-TES-103 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMR
RUO
IFFO1
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit