Learn More
Abnova™ Human HOM-TES-103 Full-length ORF (NP_542769.2, 1 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$499.00 - $769.00
Specifications
Accession Number | NP_542769.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 25900 |
Molecular Weight (g/mol) | 49.3kDa |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-947-709
|
Abnova™
H00025900P01L |
25 ug |
Each for $769.00
|
|
|||||
89-947-708
|
Abnova™
H00025900P01S |
10 ug |
Each for $499.00
|
|
|||||
Description
This gene is a member of the intermediate filament family. Intermediate filaments are proteins which are primordial components of the cytoskeleton and nuclear envelope. The proteins encoded by the members of this gene family are evolutionarily and structurally related but have limited sequence homology, with the exception of the central rod domain. Alternative splicing has been observed for this gene and three transcript variants encoding different isoforms have been identified. Other alternatively spliced transcripts may exist, but their biological validity has not been confirmed. [provided by RefSeq]
Sequence: MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMRSpecifications
NP_542769.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MGGRKRERKAAVEEDTSLSESEGPRQPDGDEEESTALSINEEMQRMLNQLREYDFEDDCDSLTWEETEETLLLWEDFSGYAMAAAEAQGEQQEDSLEKVIKDTESLFKTREKEYQETIDQIELELATAKNDMNRHLHEYMEMCSMKRGLDVQMETCRRLITQSGDRKSPAFTAVPLSDPPPPPSEAEDSDRDVSSDSSMR | |
RUO | |
IFFO1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
25900 | |
HOM-TES-103 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp586I2223/FLJ20703/HOM-TES-103/IFFO/MGC117359 | |
IFFO1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.