Learn More
Abnova Human HOXA13 Partial ORF (NP_000513, 208 a.a. - 306 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
$384.10 - $599.28
Specifications
Name | HOXA13 (Human) Recombinant Protein (Q01) |
---|---|
Recombinant | Yes |
For Use With (Application) | Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) |
Molecular Weight | 36.63kDa |
Storage Requirements | Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-947-738 |
Abnova
H00003209Q01L |
25 ug |
Each for $599.28
|
|
89-947-737 |
Abnova
H00003209Q01S |
10 ug |
Each for $384.10
|
|
Description
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Expansion of a polyalanine tract in the encoded protein can cause hand-foot-uterus syndrome, also known as hand-foot-genital syndrome. [provided by RefSeq]
Sequence: DKYMDTAGPAAEEFSSRAKEFAFYHQGYAAGPYHHHQPMPGYLDMPVVPGLGGPGESRHEPLGLPMESYQPWALPNGWNGQMYCPKEQAQPPHLWKSTLSpecifications
HOXA13 (Human) Recombinant Protein (Q01) | |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Yes | |
36.63kDa |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.