Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXA3 Partial ORF (NP_705895, 1 a.a. - 77 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003200Q01S

 View more versions of this product

Catalog No. 89-947-739


Add to cart

Description

Description

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq]

Sequence: MQKATYYDSSAIYGGYPYQAANGFAYNANQQPYPASAALGADGEYHRPACSLQSPSSAGGHPKAHELSEACLRTLSA
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.21kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MQKATYYDSSAIYGGYPYQAANGFAYNANQQPYPASAALGADGEYHRPACSLQSPSSAGGHPKAHELSEACLRTLSA
RUO
HOXA3
Wheat Germ (in vitro)
GST
NP_705895
Liquid
3200
HOXA3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOX1|HOX1E|MGC10155
HOXA3
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit