Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXA6 Partial ORF (NP_076919, 31 a.a. - 130 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name HOXA6 (Human) Recombinant Protein (Q01)
Recombinant Yes
For Use With (Application) Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Molecular Weight 36.74kDa
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-746 View Documents Abnova™
H00003203Q01S
10 ug Each for $384.10
Add to cart
 
89-947-747 View Documents Abnova™
H00003203Q01L
25 ug Each for $599.28
Add to cart
 
Description

Description

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation. [provided by RefSeq]

Sequence: GYDALRPFPASYGASSLPDKTYTSPCFYQQSNSVLACNRASYEYGASCFYSDKDLSGASPSGSGKQRGPGDYLHFSPEQQYKPDSSSGQGKALHDEGADR
Specifications

Specifications

HOXA6 (Human) Recombinant Protein (Q01)
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Yes
36.74kDa
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit