Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXC10 Partial ORF (AAH01293, 158 a.a. - 257 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003226Q01L

 View more versions of this product

Catalog No. 89-947-789


Add to cart

Description

Description

This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The protein level is controlled during cell differentiation and proliferation, which may indicate this protein has a role in origin activation. [provided by RefSeq]

Sequence: LDKTPHCSGANDFEAPFEQRASLNPRAEHLESPQLGGKVSFPETPKSDSQTPSPNEIKTEQSLAGPKGSPSESEKERAKAADSSPDTSDNEAKEEIKAEN
Specifications

Specifications

wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LDKTPHCSGANDFEAPFEQRASLNPRAEHLESPQLGGKVSFPETPKSDSQTPSPNEIKTEQSLAGPKGSPSESEKERAKAADSSPDTSDNEAKEEIKAEN
RUO
HOXC10
Wheat Germ (in vitro)
GST
AAH01293
Liquid
3226
HOXC10 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOX3I|MGC5259
HOXC10
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit