Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human HOXD10 Antibody, R&D Systems™

Goat Antibody
$200.50 - $538.00
Specifications
Antigen | HOXD10 |
---|---|
Dilution | Immunocytochemistry 5-15 ug/mL |
Applications | Immunocytochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
AF10413SP
![]() |
R&D Systems
AF10413SP |
25 μg |
Each for $200.50
|
|
|||||
AF10413100
![]() |
R&D Systems
AF10413100 |
100 μg |
Each for $538.00
|
|
|||||
Description
HOXD10 specifically detects HOXD10 in Human samples. It is validated for Immunocytochemistry.Specifications
HOXD10 | |
Immunocytochemistry | |
Unconjugated | |
Goat | |
Human | |
P28358 | |
3236 | |
IgG | |
Antigen Affinity-purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Immunocytochemistry 5-15 ug/mL | |
Polyclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
homeo box 4D, homeo box D10, homeobox D10, Homeobox protein Hox-4D, Homeobox protein Hox-4E, homeobox protein Hox-D10, HOX4, Hox-4.4, HOX4D, HOX4DHOX4, HOX4E | |
E. coli-derived recombinant human HOXD10, Arg153-Thr262, Accession # P28358 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title