Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HOXD10 Partial ORF (NP_002139.2, 44 a.a. - 143 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00003236Q01S

 View more versions of this product

Catalog No. 89-947-811


Add to cart

Description

Description

This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm's tumor and congenital vertical talus (also known as “rocker-bottom foot” deformity or congenital convex pes valgus) and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease. [provided by RefSeq]

Sequence: GTYGMQTCGLLPSLAKREVNHQNMGMNVHPYIPQVDSWTDPNRSCRIEQPVTQQVPTCSFTTNIKEESNCCMYSDKRNKLISAEVPSYQRLVPESCPVEN
Specifications

Specifications

NP_002139.2
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
GTYGMQTCGLLPSLAKREVNHQNMGMNVHPYIPQVDSWTDPNRSCRIEQPVTQQVPTCSFTTNIKEESNCCMYSDKRNKLISAEVPSYQRLVPESCPVEN
RUO
HOXD10
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
3236
HOXD10 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOX4|HOX4D|HOX4E|Hox-4.4
HOXD10
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit