Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HPS1 Full-length ORF (NP_872577.1, 1 a.a. - 324 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name HPS1 (Human) Recombinant Protein (P02)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 62.9kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-850 View Documents Abnova™
H00003257P02L
25 ug Each for $599.28
Add to cart
 
89-947-849 View Documents Abnova™
H00003257P02S
10 ug Each for $384.10
Add to cart
 
Description

Description

This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. The encoded protein is a component of three different protein complexes termed biogenesis of lysosome-related organelles complex (BLOC)-3, BLOC4, and BLOC5. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 1. Multiple transcript variants encoding distinct isoforms have been identified for this gene; the full-length sequences of some of these have not been determined yet. [provided by RefSeq]

Sequence: MKCVLVATEGAEVLFYWTDQEFEESLRLKFGQSENEEEELPALEDQLSTLLAPVIISSMTMLEKLSDTYTCFSTENGNFLYVLHLFGECLFIAINGDHTESEGDLRRKLYVLKYLFEVHFGLVTVDGHLIRKELRPPDLAQRVQLWEHFQSLLWTYSRLREQEQCFAVEALERLIHPQLCELCIEALERHVIQAVNTSPERGGEEALHAFLLVHSKLLAFYSSHSASSLRPADLLALILLVQDLYPSESTAEDDIQPSPRRARSSQNIPVQQAWSPHSTGPTGGSSAETETDSFSLPEEYFTPAPSPGDQSSGEDRRKAGGNNS
Specifications

Specifications

HPS1 (Human) Recombinant Protein (P02)
Glutathione Sepharose 4 Fast Flow
62.9kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit