Learn More
Abnova™ Human HPS5 Full-length ORF (AAH33640.1, 1 a.a. - 1015 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00011234P01L
Description
This gene encodes a protein that may play a role in organelle biogenesis associated with melanosomes, platelet dense granules, and lysosomes. This protein interacts with Hermansky-Pudlak syndrome 6 protein and may interact with the cytoplasmic domain of integrin, alpha-3. Mutations in this gene are associated with Hermansky-Pudlak syndrome type 5. Multiple transcript variants encoding two distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MYVSSEHKGRRVTALCWDTAILRVFVGDHAGKVSAIKLNTSKQAKAAAAFVMFPVQTITTVDSCVVQLDYLDGRLLISSLTRSFLCDTEREKFWKIGNKERDGEYGACFFPGRCSGGQQPLIYCARPGSRMWEVNFDGEVISTHQFKKLLSLPPLPVITLRSEPQYDHTAGSSQSLSFPKLLHLSEHCVLTWTERGIYIFIPQNVQVLLWSEVKDIQDVAVCRNELFCLHLNGKVSHLSLISVERCVERLLRRGLWNLAARTCCLFQNSVIASRARKTLTADKLEHLKSQLDHGTYNDLISQLEELILKFEPLDSACSSRRSSISSHESFSILDSGIYRIISSRRGSQSDEDSCSLHSQTLSEDERFKEFTSQQEEDLPDQCCGSHGNEDNVSHAPVMFETDKNETFLPFGIPLPFRSPSPLVSLQAVKESVSSFVRKTTEKIGTLHTSPDLKVRPELRGDEQSCEEDVSSDTCPKEEDTEEEKEVTSPPPEEDRFQELKVATAEAMTKLQDPLVLFESESLRMVLQEWLSHLEKTFAMKDFSGVSDTDNSSMKLNQDVLLVNESKKGILDEDNEKEKRDSLGNEESVDKTACECVRSPRESLDDLFQICSPCAIASGLRNDLAELTTLCLELNVLNSKIKSTSGHVDHTLQQYSPEILACQFLKKYFFLLNLKRAKESIKLSYSNSPSVWDTFIEGLKEMASSNPVYMEMEKGDLPTRLKLLDDEVPFDSPLLVVYATRLYEKFGESALRSLIKFFPSILPSDIIQLCHHHPAEFLAYLDSLVKSRPEDQRSSFLESLLQPESLRLDWLLLAVSLDAPPSTSTMDDEGYPRPHSHLLSWGYSQLILHLIKLPADFITKEKMTDICRSCGFWPGYLILCLELERRREAFTNIVYLNDMSLMKGDNGWIPETVEEWKLLLHLIQSKSTRPAPQESLNGSLSDGPSPINVENVALLLAKAMGPDRAWSLLQECGLALELSEKFTRTCDILRIAEKRQRALIQSMLEKCDRFLWSQQASpecifications
AAH33640.1 | |
Liquid | |
11234 | |
HPS5 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MYVSSEHKGRRVTALCWDTAILRVFVGDHAGKVSAIKLNTSKQAKAAAAFVMFPVQTITTVDSCVVQLDYLDGRLLISSLTRSFLCDTEREKFWKIGNKERDGEYGACFFPGRCSGGQQPLIYCARPGSRMWEVNFDGEVISTHQFKKLLSLPPLPVITLRSEPQYDHTAGSSQSLSFPKLLHLSEHCVLTWTERGIYIFIPQNVQVLLWSEVKDIQDVAVCRNELFCLHLNGKVSHLSLISVERCVERLLRRGLWNLAARTCCLFQNSVIASRARKTLTADKLEHLKSQLDHGTYNDLISQLEELILKFEPLDSACSSRRSSISSHESFSILDSGIYRIISSRRGSQSDEDSCSLHSQTLSEDERFKEFTSQQEEDLPDQCCGSHGNEDNVSHAPVMFETDKNETFLPFGIPLPFRSPSPLVSLQAVKESVSSFVRKTTEKIGTLHTSPDLKVRPELRGDEQSCEEDVSSDTCPKEEDTEEEKEVTSPPPEEDRFQELKVATAEAMTKLQDPLVLFESESLRMVLQEWLSHLEKTFAMKDFSGVSDTDNSSMKLNQDVLLVNESKKGILDEDNEKEKRDSLGNEESVDKTACECVRSPRESLDDLFQICSPCAIASGLRNDLAELTTLCLELNVLNSKIKSTSGHVDHTLQQYSPEILACQFLKKYFFLLNLKRAKESIKLSYSNSPSVWDTFIEGLKEMASSNPVYMEMEKGDLPTRLKLLDDEVPFDSPLLVVYATRLYEKFGESALRSLIKFFPSILPSDIIQLCHHHPAEFLAYLDSLVKSRPEDQRSSFLESLLQPESLRLDWLLLAVSLDAPPSTSTMDDEGYPRPHSHLLSWGYSQLILHLIKLPADFITKEKMTDICRSCGFWPGYLILCLELERRREAFTNIVYLNDMSLMKGDNGWIPETVEEWKLLLHLIQSKSTRPAPQESLNGSLSDGPSPINVENVALLLAKAMGPDRAWSLLQECGLALELSEKFTRTCDILRIAEKRQRALIQSMLEKCDRFLWSQQA | |
RUO | |
HPS5 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
141.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AIBP63/KIAA1017 | |
HPS5 | |
Yes | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.