Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HRBL Full-length ORF (ENSP00000262935, 1 a.a. - 155 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

$384.10 - $599.28

Specifications

Name HRBL (Human) Recombinant Protein (P01)
Recombinant Yes
Purification Method Glutathione Sepharose 4 Fast Flow
For Use With (Application) Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Molecular Weight 43.6kDa
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
89-947-881 View Documents Abnova™
H00003268P01L
25 ug Each for $599.28
Add to cart
 
89-947-880 View Documents Abnova™
H00003268P01S
10 ug Each for $384.10
Add to cart
 
Description

Description

This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. The 3' UTR of this gene contains an insulin receptor substrate 3-like pseudogene. [provided by RefSeq]

Sequence: MVMAAKKGPGPGGGVSGGKAEAEAASEVWCRRVRELGGCSQAGNRHCFECAQRGVTYVDITVGSFVCTTCSGLLRGLNPPHRVKSISMTTFTEPEVVFLQSRGNEVCRKIWLGLFDARTSLVPDSRDPQKVKEFLQEKYEKKRWPDTFPRRLCQL
Specifications

Specifications

HRBL (Human) Recombinant Protein (P01)
Glutathione Sepharose 4 Fast Flow
43.6kDa
Yes
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit