Learn More
Abnova™ Human HRBL Partial ORF (NP_006067, 131 a.a. - 228 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00003268Q01S
Description
This gene is a member of the HIV-1 Rev binding protein (HRB) family and encodes a protein with one Arf-GAP zinc finger domain, several phe-gly (FG) motifs, and four asn-pro-phe (NPF) motifs. This protein interacts with Eps15 homology (EH) domains and plays a role in the Rev export pathway, which mediates the nucleocytoplasmic transfer of proteins and RNAs. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. The 3' UTR of this gene contains an insulin receptor substrate 3-like pseudogene. [provided by RefSeq]
Sequence: VKEFLQEKYEKKRWYVPPDQVKGPTYTKGSASTPVQGSIPEGKPLRTLLGDPAPSLSVAASTSSQPVSQSHARTSQARSTQPPPHSSVKKASTDLLADSpecifications
NP_006067 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
VKEFLQEKYEKKRWYVPPDQVKGPTYTKGSASTPVQGSIPEGKPLRTLLGDPAPSLSVAASTSSQPVSQSHARTSQARSTQPPPHSSVKKASTDLLAD | |
RUO | |
AGFG2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3268 | |
HRBL (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HRBL/RABR | |
AGFG2 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.