Learn More
Abnova™ Human HS2ST1 Partial ORF (NP_036394, 130 a.a. - 229 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Supplier: Abnova™ H00009653Q01S
Description
Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. This gene encodes a member of the heparan sulfate biosynthetic enzyme family that transfers sulfate to the 2 position of the iduronic acid residue of heparan sulfate. The disruption of this gene resulted in no kidney formation in knockout embryonic mice, indicating that the absence of this enzyme may interfere with the signaling required for kidney formation. Two alternatively spliced transcript variants that encode different proteins have been found for this gene. [provided by RefSeq]
Sequence: SWKEMKPGFYHGHVSYLDFAKFGVKKKPIYINVIRDPIERLVSYYYFLRFGDDYRPGLRRRKQGDKKTFDECVAEGGSDCAPEKLWLQIPFFCGHSSECWSpecifications
NP_036394 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
SWKEMKPGFYHGHVSYLDFAKFGVKKKPIYINVIRDPIERLVSYYYFLRFGDDYRPGLRRRKQGDKKTFDECVAEGGSDCAPEKLWLQIPFFCGHSSECW | |
RUO | |
HS2ST1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
9653 | |
HS2ST1 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ11317/KIAA0448/MGC131986/dJ604K5.2 | |
HS2ST1 | |
Recombinant | |
wheat germ expression system |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.