Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HS3ST3A1 Partial ORF (NP_006033, 308 a.a. - 406 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00009955Q01L

 View more versions of this product

Catalog No. 89-947-915


Add to cart

Description

Description

Heparan sulfate biosynthetic enzymes are key components in generating a myriad of distinct heparan sulfate fine structures that carry out multiple biologic activities. The enzyme encoded by this gene is a member of the heparan sulfate biosynthetic enzyme family. It is a type II integral membrane protein and possesses heparan sulfate glucosaminyl 3-O-sulfotransferase activity. The sulfotransferase domain of this enzyme is highly similar to the same domain of heparan sulfate D-glucosaminyl 3-O-sulfotransferase 3A1, and these two enzymes sulfate an identical disaccharide. This gene is widely expressed, with the most abundant expression in liver and placenta. [provided by RefSeq]

Sequence: LFVSGERLISDPAGELGRVQDFLGLKRIITDKHFYFNKTKGFPCLKKAEGSSRPHCLGKTKGRTHPEIDREVVRRLREFYRPFNLKFYQMTGHDFGWDG
Specifications

Specifications

NP_006033
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LFVSGERLISDPAGELGRVQDFLGLKRIITDKHFYFNKTKGFPCLKKAEGSSRPHCLGKTKGRTHPEIDREVVRRLREFYRPFNLKFYQMTGHDFGWDG
RUO
HS3ST3A1
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
9955
HS3ST3A1 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
30ST3A1|3OST3A1
HS3ST3A1
Yes
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit