Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human HSD17B12 Full-length ORF (AAH12536.1, 1 a.a. - 98 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Manufacturer:  Abnova™ H00051144P01S

 View more versions of this product

Catalog No. 89-947-946


Add to cart

Description

Description

The enzyme 17-beta hydroxysteroid dehydrogenase-12 (HSD17B12) uses NADPH to reduce 3-ketoacyl-CoA to 3-hydroxyacyl-CoA during the second step of fatty acid elongation.[supplied by OMIM]

Sequence: MESALPAAGFLYWVGAGTVAYLALRISYSLFTALRVWGVGNEAGVGPGLGEWAVVTGSTDGIGKSYAEELAKHGMKVVLISRSKDKLDQVSSEISNYT
Specifications

Specifications

AAH12536.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KAR|SDR12C1
HSD17B12
Yes
wheat germ expression system
Liquid
51144
HSD17B12 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MESALPAAGFLYWVGAGTVAYLALRISYSLFTALRVWGVGNEAGVGPGLGEWAVVTGSTDGIGKSYAEELAKHGMKVVLISRSKDKLDQVSSEISNYT
RUO
HSD17B12
Wheat Germ (in vitro)
GST
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit