Learn More
Novus Biologicals™ Human HSF1 ELISA Kit (Colorimetric)
ELISA kits are ideal for the quantification of target antigens in tissue extracts, serum and cell culture.
Manufacturer:  Novus Biologicals™ NBP262149
Catalog No. NBP262149
Add to cart
Description
Description
The Human HSF1 ELISA Kit (Colorimetric) measures human HSF1 in cell lysates and tissue.
Specifications
Specifications
1 Kit
0.39 - 12.5 ng/ml
3297
35 pg/ml
HSF1
HSF1
Cell Lysates, Tissue
ELISA
ELISA Kit (Colorimetric)
Storage of components varies. See protocol for specific instructions.
Human